Learn More
ABCB6 Rabbit anti-Dog, Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ABCB6.
Brand: Abnova PAB9926.100ug
Description
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This half-transporter likely plays a role in mitochondrial function. Localized to 2q26, this gene is considered a candidate gene for lethal neonatal metabolic syndrome, a disorder of mitochondrial function. [provided by RefSeq
Specifications
| ABCB6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ABCB6. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Q9NP58 | |
| ABCB6 | |
| A synthetic peptide corresponding to amino acids 440-45 of human ABCB6. | |
| RUO | |
| 10058 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:40000-1:200000) Western Blot (1:1000-1:5000) The optimal working dilution should be determined by the end user. | |
| ABCB6 | |
| ABC/ABC14/EST45597/FLJ22414/MTABC3/PRP/umat | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Canine, Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only