Learn More
ABCB5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ABCB5.
Brand: Abnova PAB9925.100ug
Description
ABCB5 belongs to the ATP-binding cassette (ABC) transporter superfamily of integral membrane proteins. These proteins participate in ATP-dependent transmembrane transport of structurally diverse molecules ranging from small ions, sugars, and peptides to more complex organic molecules (Chen et al., 2005 [PubMed 15760339]).[supplied by OMIM
Specifications
| ABCB5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ABCB5. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| AAW31630;Q515Q7 | |
| ABCB5 | |
| A synthetic peptide corresponding to internal region of human ABCB5. | |
| RUO | |
| 340273 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:80000-1:85000) Western Blot (1:10000) The optimal working dilution should be determined by the end user. | |
| ABCB5 | |
| ABCB5alpha/ABCB5beta/EST422562 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only