missing translation for 'onlineSavingsMsg'
Learn More

RAB9B Antibody (3C9), Novus Biologicals™

Product Code. 18315659 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18315659 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18315659 Supplier Novus Biologicals Supplier No. H00051209M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RAB9B Monoclonal antibody specifically detects RAB9B in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB9B
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3C9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_057454
Gene Alias RAB9B, member RAS oncogene family, Rab-9L, RAB9-like protein, RAB9LRab-9-like protein, ras-related protein Rab-9B
Host Species Mouse
Immunogen RAB9B (NP_057454, 102 a.a. ~ 201 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QKEFIYYADVKDPEHFPFVVLGNKVDKEDRQVTTEEAQTWCMENGDYPYLETSAKDDTNVTVAFEEAVRQVLAVEEQLEHCMLGHTIDLNSGSKAGSSCC
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51209
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.