missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB40A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93889-0.1ml
This item is not returnable.
View return policy
Description
RAB40A Polyclonal antibody specifically detects RAB40A in Mouse, Rat samples. It is validated for Western Blot
Specifications
| RAB40A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC142061, Protein Rar-2, RAB40A, member RAS oncogene family, Rar-2, RAR2, RAR2A, ras-related protein Rab-40A, SOCS box containing protein RAR2A, SOCS box-containing protein RAR2A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human RAB40A (NP_543155.2). MSAPGSPDQAYDFLLKFLLVGDRDVGKSEILESLQDGAAESPYSHLGGIDYKTTTILLDGQRVKLKLWDTSGQGRFCTIFRSYSRGAQGVILVYDIANRWSFEGMDRWIKKIEEHAPGVPKILVGNRLHLAFKRQVPREQAQAYAERLGV | |
| 0.1 mL | |
| Signal Transduction | |
| 142684 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction