Learn More
(R)-Ostarine, TRC
Product Code. 30009546
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009546 25 mg 25mg
30008196 50 mg 50mg
30009068 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009546 Supplier Toronto Research Chemicals Supplier No. TRCO70350525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1132656-73-5
Molecular Formula C19 H14 F3 N3 O3
Molecular Weight (g/mol) 389.33
Synonym Propanamide, 3-(4-cyanophenoxy)-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methyl-, (2R)- (ACI), (2R)-3-(4-Cyanophenoxy)-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide (ACI), (R)-N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-(4-cyanophenoxy)-2-hydroxy-2-methylpropanamide, (R)-Enobosarm, (R)-Ostarine
IUPAC Name (2R)-3-(4-cyanophenoxy)-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide
SMILES C[C@@](O)(COc1ccc(cc1)C#N)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C19H14F3N3O3/c1-18(27,11-28-15-6-2-12(9-23)3-7-15)17(26)25-14-5-4-13(10-24)16(8-14)19(20,21)22/h2-8,27H,11H2,1H3,(H,25,26)/t18-/m1/s1
Formula Weight 389.0987
Percent Purity >95%
Chemical Name or Material (R)-Ostarine
Grade HPLC