Learn More
Ganciclovir Mono-O-acetate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Ganciclovir Mono-O-acetate
Specifications
Specifications
| Chemical Name or Material | Ganciclovir Mono-O-acetate |
| CAS | 88110-89-8 |
| Analyte or Component Names | Ganciclovir Mono-O-acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H15 N5 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 6H-Purin-6-one, 9-[[2-(acetyloxy)-1-(hydroxymethyl)ethoxy]methyl]-2-amino-1,9-dihydro-, 2-[(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-3-acetoxy-1-propanol, 2-[(2-amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]-3-hydroxypropyl acetate, Ganciclovir Mono-O-acetate, Monoacetoxyganciclovir, O-Acetoxyganciclovir |
| InChI Formula | InChI=1S/C11H15N5O5/c1-6(18)20-3-7(2-17)21-5-16-4-13-8-9(16)14-11(12)15-10(8)19/h4,7,17H,2-3,5H2,1H3,(H3,12,14,15,19) |
| SMILES | CC(=O)OCC(CO)OCn1cnc2C(=O)NC(=Nc12)N |
| IUPAC Name | [2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]-3-hydroxypropyl] acetate |
| Show More |