missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Thrombospondin-1 Protein, CF

Product Code. 18106150
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18106150

Brand: R&D Systems™ 3074TH050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by the ability of the immobilized protein to support the adhesion of SVEC4-10 mouse vascular endothelial cells. When 2 x 104 cells/well are added to DTT treated THBS-1 coated plates (10 μg/mL, 100 μL/well), > 50% will adhere after 1 hour at 37 °C.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07996-1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in TBS, NaCl, and Trehalose.
Gene ID (Entrez) 7057
Molecular Weight (g/mol) M.W. (Observed): 135 - 165 kDa, reducing conditions; M.W. (theoretical): 129 kDa
Name Thrombospondin-1
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Thrombospondin-1 protein Asn19-Pro1170, with Thr523Ala substitution and a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol THBS1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Thrombospondin-1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.