missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human B4GalT1 Protein, CF
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
Measured by its ability to transfer galactose from UDP-galactose to N-Acetyl-alpha-D-glucosamine. The specific activity is >2,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | NP_001488 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 2683 |
| Molecular Weight (g/mol) | M.W. (Observed): 45-55 kDa; M.W. (theoretical): 40 kDa |
| Name | beta-1,4-Galactosyltransferase 1/B4GalT1 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived human beta-1,4-Galactosyltransferase 1/B4GalT1 protein Gly44-Ser398, with an N-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction