missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-4-(2-Methyl-3,3-diphenylpropyl)morpholine ((1R,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate, TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | (R)-4-(2-Methyl-3,3-diphenylpropyl)morpholine ((1R,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate |
| Analyte or Component Names | (R)-4-(2-Methyl-3,3-diphenylpropyl)morpholine ((1R,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20H25NO . C10H16O4S |
| Synonym | (R)-4-(2-Methyl-3,3-diphenylpropyl)morpholine (R)-10-Camphorsulfonic Acid Salt |
| Solution Type | Neat |
| SMILES | [H]C(C1=CC=CC=C1)(C2=CC=CC=C2)[C@@H](C)CN3CCOCC3.CC4(C)[C@@H]5CC[C@]4(CS(=O)(O)=O)C(C5)=O |
| Industry Type | Forensics |
| Molecular Weight (g/mol) | 527.72 |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction