missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinapril Benzyl Ester Maleate, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Quinapril Benzyl Ester Maleate
Specifications
Specifications
| Chemical Name or Material | Quinapril Benzyl Ester Maleate |
| CAS | 82586-54-7 |
| Analyte or Component Names | Quinapril Benzyl Ester Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C32 H36 N2 O5 . C4 H4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Quinapril Imp. F (Pharmeuropa) as Maleate: Benzyl (3S)-2-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenyl]amino]propanoyl]-1,2,3,4-tetra-hydroisoquinoline-3-carboxylate Maleate |
| InChI Formula | InChI=1S/C32H36N2O5.C4H4O4/c1-3-38-31(36)28(19-18-24-12-6-4-7-13-24)33-23(2)30(35)34-21-27-17-11-10-16-26(27)20-29(34)32(37)39-22-25-14-8-5-9-15-25;5-3(6)1-2-4(7)8/h4-17,23,28-29,33H,3,18-22H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t23-,28-,29-;/m0./s1 |
| SMILES | CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2Cc3ccccc3C[C@H]2C(=O)OCc4ccccc4.OC(=O)\C=C/C(=O)O |
| IUPAC Name | benzyl (3S)-2-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylate;(Z)-but-2-enedioic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction