Learn More
Applied Biosystems™ QuantStudio™ 7 Pro Real-Time PCR System Extended Warranty Package
Product Code. 17112126 Shop All Applied Biosystems Products
Change view
Click to view available options
Format:
96-well Plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
17112126 96-well Plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17112126 Supplier Applied Biosystems™ Supplier No. A47205
 Restricted Item
Estimated Shipment: 24-11-2026
Adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint.
  • Includes facial authentication, voice commands, Smart Help, multiplex capability, motorized interchangeable blocks, simplified workflow, intuitive software, and large capacitive touch-screen interface.
  • Provides reproducibility with minimal well-to-well and instrument-to-instrument variation.
  • Designed for ultimate ease-of-use, the system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab.
  • Easy and convenient hands-free operation with facial authentication and voice commands
  • Access, analyze, and share data anytime, anywhere with our Connect cloud-based platform for real-time information access, secure data storage, and online results sharing
  • The Connect platform provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the platform are web browser-based software tools that are flexible, fast, and easy to use. These software tools support a variety of analyses including relative quantitation, standard curve, high resolution melt (HRM), genotyping, and presence/absence. They are compatible with Mac or PC as well as most Applied Biosystems instrument file types
  • The system is delivered with one block type; other block types may be purchased separately.
  • In addition, an Orbitor Microplate Mover is available for this system to support high-throughput workflows.
  • Run hundreds of real-time PCR reactions using TaqMan Array 384-well microfluidic cards. And when integrated with the Orbitor Microplate Mover (available separately), the QuantStudio 7 Pro system maximizes your throughput capabilities.
  • Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex system, which features six decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.
  • Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
TRUSTED_SUSTAINABILITY
Specifications
Block Format Interchangeable
Format 96-well Plate
For Use With (Equipment) QuantStudio™, ViiA™ 7 System
Capacity 0.1 mL
Product Line QuantStudio
Display Type Touchscreen
Shipping Condition Room Temperature