Learn More
MP Biomedicals™ qPCR& Go Probe Lo-ROX Kit
Fast and sensitive qPCR Probe Master Mix. For most Agilent platforms and some newer ABI platforms.
397.50€ - 1371.64€
Specifications
Content And Storage | dNTPs, stabilizers and enhancers. Store at -20°C |
---|---|
Description | Fast and sensitive qPCR Probe Master Mix. For most Agilent platforms and some newer ABI platforms. |
Product Type | qPCR Kit |
For Use With (Application) | Real Time PCR (qPCR) |
Concentration | 2x |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15828551
|
MP Biomedicals™
11EBI05050 |
5 x 1 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15838551
|
MP Biomedicals™
11EBI05200 |
4 x 5 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- The qPCR& Go Probe Lo-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
- The kit has been formulated for use with probe-detection technology, including TaqMan™, Scorpions™ and molecular beacon probes.
- A combination of the latest advances in buffer chemistry and PCR enhancers, together with a hot-start DNA polymerase, ensures that the qPCR& Go Probe Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
- qPCR& Go Probe is provided as a 2x mastermix containing all the components necessary for real-time PCR, including dNTPs, stabilizers and enhancers.
Specifications
dNTPs, stabilizers and enhancers. Store at -20°C | |
qPCR Kit | |
2x |
Fast and sensitive qPCR Probe Master Mix. For most Agilent platforms and some newer ABI platforms. | |
Real Time PCR (qPCR) | |
500 |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.