missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ qPCR& Go Probe Lo-ROX Kit

Fast and sensitive qPCR Probe Master Mix. For most Agilent platforms and some newer ABI platforms.

Brand:  MP Biomedicals™ 11EBI05200

Additional Details : Weight : 0.00500kg

 View more versions of this product

Product Code. 15838551

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

  • The qPCR& Go Probe Lo-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
  • The kit has been formulated for use with probe-detection technology, including TaqMan™, Scorpions™ and molecular beacon probes.
  • A combination of the latest advances in buffer chemistry and PCR enhancers, together with a hot-start DNA polymerase, ensures that the qPCR& Go Probe Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
  • qPCR& Go Probe is provided as a 2x mastermix containing all the components necessary for real-time PCR, including dNTPs, stabilizers and enhancers.
Specifications

Specifications

dNTPs, stabilizers and enhancers. Store at -20°C
qPCR Kit
Real Time PCR (qPCR)
500
Fast and sensitive qPCR Probe Master Mix. For most Agilent platforms and some newer ABI platforms.
4 x 5 mL
2x
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.