missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2,3-dicarboxylic Acid Monohydrate, TRC

Product Code. 30425137
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425137 50 mg 50mg
30410342 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30425137 Supplier Toronto Research Chemicals Supplier No. TRCP99788050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1329802-41-6
Molecular Formula C6H7NO5
Molecular Weight (g/mol) 173.12
Synonym 1H-Pyrrole-2,3-dicarboxylic Acid Hydrate, PDCA
IUPAC Name 1H-pyrrole-2,3-dicarboxylic acid;hydrate
SMILES C1=CNC(=C1C(=O)O)C(=O)O.O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H5NO4.H2O/c8-5(9)3-1-2-7-4(3)6(10)11;/h1-2,7H,(H,8,9)(H,10,11);1H2
Formula Weight 173.0324
Percent Purity >95%
Chemical Name or Material Pyrrole-2,3-dicarboxylic Acid Monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.