CAS RN 1329802-41-6
CAS RN 1329802-41-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pyrrole-2,3-dicarboxylic Acid Monohydrate, TRC
CAS: 1329802-41-6 Molecular Formula: C6H7NO5 Molecular Weight (g/mol): 173.12 Synonym: 1H-Pyrrole-2,3-dicarboxylic Acid Hydrate,PDCA IUPAC Name: 1H-pyrrole-2,3-dicarboxylic acid;hydrate SMILES: C1=CNC(=C1C(=O)O)C(=O)O.O