Learn More
Pyronin Y, 0.2% w/v aq. soln., Thermo Scientific™
Product Code. 15433279
Change view
Click to view available options
Quantity:
10 mL
25 mL
Unit Size:
10mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433279 10 mL 10mL
15443279 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15433279 Supplier Thermo Scientific Alfa Aesar Supplier No. J60505.AB
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 92-32-0
Molecular Formula C17H19ClN2O
Molecular Weight (g/mol) 302.802
MDL Number MFCD00011725
InChI Key INCIMLINXXICKS-UHFFFAOYSA-M
Synonym C.I. 45005; Pyronin G
PubChem CID 7085
ChEBI CHEBI:87347
IUPAC Name [6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC2=C(C=C1)C=C3C=CC(=[N+](C)C)C=C3O2.[Cl-]
Specifications
Concentration 0.2% w/v aq. soln.
Beilstein 3795789
Merck Index 14,8007
Formula Weight 302.8
Odor Odorless
Color Pink
Quantity 10 mL
Physical Form Liquid
Chemical Name or Material Pyronin Y