missing translation for 'onlineSavingsMsg'
Learn More

Pyrimethanil-4'-hydroxy 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30314426
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30314426

Brand: Dr. Ehrenstorfer DREA16658515AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrimethanil-4'-hydroxy
CAS 81261-84-9
Analyte or Component Names Pyrimethanil-4'-hydroxy
Shipping Condition +20°C
Molecular Formula C12 H13 N3 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Phenol, 4-[(4,6-dimethyl-2-pyrimidinyl)amino]-, 4-[(4,6-Dimethyl-2-pyrimidinyl)amino]phenol, 4'-Hydroxy Pyrimethanil
InChI Formula InChI=1S/C12H13N3O/c1-8-7-9(2)14-12(13-8)15-10-3-5-11(16)6-4-10/h3-7,16H,1-2H3,(H,13,14,15)
SMILES Cc1cc(C)nc(Nc2ccc(O)cc2)n1
IUPAC Name 4-[(4,6-dimethylpyrimidin-2-yl)amino]phenol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 215.25
Quantity 1 mL
Formula Weight 215.1059 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.