Learn More
Pyrex™ Aspirator Bottle with Ground Glass Side Socket
Product Code. 13421909
Click to view available options
Capacity (Metric):
10,000 mL
20,000 mL
2000 mL
5000 mL
Unit Size:
Each
Pack of 5
Product Code. 13421909
Brand: Pyrex 1541/12D
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Manufactured from borosilicate glass
  • Ground side socket (size 29/32) to accept stopcock assemblies
  • Unground top neck to accept rubber stoppers
Compliance
ISO 4796
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2000 mL
Diameter (Metric) Inner Neck 28 mm
Material Borosilicate Glass
Height (Metric) 248 mm
Product Type Aspirator Bottle
Diameter (Metric) 138 mm