Learn More
Pyrex™ Aspirator Bottle with Ground Glass Side Socket
Product Code. 11377884
Change view
Click to view available options
Capacity (Metric):
10,000 mL
20,000 mL
2000 mL
5000 mL
Unit Size:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11377884 20,000 mL Each
11357884 10,000 mL Each
13421909 2000 mL Pack of 5
11347884 5000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11377884 Supplier Pyrex Supplier No. 1541/22D
  • ONLINE EXCLUSIVE PRICE
    510.30€
    425.86€ / Pack of 1
    Save 84.44€
    16% Off
Estimated Shipment: 27-01-2027
Manufactured from borosilicate glass
  • Ground side socket (size 29/32) to accept stopcock assemblies
  • Unground top neck to accept rubber stoppers
Compliance
ISO 4796
Specifications
Capacity (Metric) 20,000 mL
Diameter (Metric) Inner Neck 59 mm
Material Borosilicate Glass
Height (Metric) 492 mm
Diameter (Metric) 288 mm
Product Type Aspirator Bottle