missing translation for 'onlineSavingsMsg'
Learn More

Pyrantel Embonate, Mikromol™

Product Code. 17992636
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992636 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17992636 Supplier Mikromol Supplier No. MM0449.00
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pyrantel Embonate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrantel pamoate
CAS 22204-24-6
Analyte or Component Names Pyrantel pamoate
Shipping Condition Room Temperature
Molecular Formula C23 H16 O6 . C11 H14 N2 S
Certifications/Compliance ISO 17034
Synonym Pyrantel embonate, 1-Methyl-2-[(E)-2-(thiophen-2-yl)ethenyl]-1,4,5,6-tetrahydropyrimidine hydrogen 4,4'-methylenebis(3-hydroxynaphthalene-2-carboxylate), 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 1,4,5,6-tetrahydro-1-methyl-2-[(1E)-2-(2-thienyl)ethenyl]pyrimidine (1:1), 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with (E)-1,4,5,6-tetrahydro-1-methyl-2-[2-(2-thienyl)ethenyl]pyrimidine (1:1), 2-Naphthoic acid, 4,4'-methylenebis[3-hydroxy-, compd. with (E)-1,4,5,6-tetrahydro-1-methyl-2-[2-(2-thienyl)vinyl]pyrimidine (1:1) (8CI), Pyrimidine, 1,4,5,6-tetrahydro-1-methyl-2-[(1E)-2-(2-thienyl)ethenyl]-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1) (9CI), Pyrimidine, 1,4,5,6-tetrahydro-1-methyl-2-[2-(2-thienyl)ethenyl]-, (E)-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1), Antiminth, CP 10423-16, Cobantril, Combantrin, Early Bird, Embovin, Helmex, Helmintox, Nemocid, Piranver, Pyrantel embonate, Pyrantel pamoate
InChI Formula InChI=1S/C23H16O6.C11H14N2S/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-13-8-3-7-12-11(13)6-5-10-4-2-9-14-10/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2,4-6,9H,3,7-8H2,1H3/b;6-5+
SMILES CN1CCCN=C1\C=C\c2cccs2.OC(=O)c3cc4ccccc4c(Cc5c(O)c(cc6ccccc56)C(=O)O)c3O
IUPAC Name 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid;1-methyl-2-[(E)-2-thiophen-2-ylethenyl]-5,6-dihydro-4H-pyrimidine
Molecular Weight (g/mol) 594.68
Quantity 250 mg
Formula Weight 594.1825 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.