missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYCR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22150-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an enzyme that catalyzes the NAD(P)H-dependent conversion of pyrroline-5-carboxylate to proline. This enzyme may also play a physiologic role in the generation of NADP(+) in some cell types. The protein forms a homopolymer and localizes to the mitochondrion. Alternate splicing results in two transcript variants encoding different isoforms.Specifications
| PYCR1 | |
| Polyclonal | |
| Unconjugated | |
| PYCR1 | |
| P5C, P5C reductase 1, P5CR, P5CR 1, PIG45, PP222, PYCR, PYCR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 209027, 287877, 5831 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P32322, Q922W5 | |
| Pycr1 | |
| PYCR1 Fusion Protein Ag17745 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction