missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PUS7 Polyclonal antibody specifically detects PUS7 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PUS7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Tris-citrate/phosphate pH 7 -8. |
| Gene Alias | EC 5.4.99.-, FLJ20485, KIAA1897, MGC17720, pseudouridylate synthase 7 homolog, pseudouridylate synthase 7 homolog (S. cerevisiae) |
| Host Species | Rabbit |
| Immunogen | Between 100 and 150 |
| Purification Method | Affinity purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?