missing translation for 'onlineSavingsMsg'
Learn More

PTPRH Antibody, Novus Biologicals™

Product Code. 18265972 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18265972 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18265972 Supplier Novus Biologicals Supplier No. NBP169289

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTPRH Polyclonal specifically detects PTPRH in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRH
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9HD43
Gene Alias EC 3.1.3.48, FLJ39938, MGC133058, MGC133059, protein tyrosine phosphatase, receptor type, H, receptor-type tyrosine-protein phosphatase H, R-PTP-H, SAP1, SAP-1, Stomach cancer-associated protein tyrosine phosphatase 1, Transmembrane-type protein-tyrosine phosphatase type H
Gene Symbols PTPRH
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PTPRH(protein tyrosine phosphatase, receptor type, H) The peptide sequence was selected from the middle region of PTPRH. Peptide sequence QTKNSVMLWWKAPGDPHSQLYVYWVQWASKGHPRRGQDPQANWVNQTSRT.
Molecular Weight of Antigen 120 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5794
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.