missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN3 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18412AF405
563.55 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
PTPN3 Polyclonal antibody specifically detects PTPN3 in Human samples. It is validated for Western Blot
Specifications
| PTPN3 | |
| Polyclonal | |
| Western Blot | |
| DKFZp686N0569, EC 3.1.3.48, protein tyrosin phosphatase H1, protein tyrosine phosphatase, non-receptor type 3, Protein-tyrosine phosphatase H1, PTP-H1, PTPH1cytoskeletal-associated protein tyrosine phosphatase, tyrosine phosphatase, tyrosine-protein phosphatase non-receptor type 3 | |
| Recognizes region between residues Between between 175 and 225.. | |
| 0.1 mL | |
| Immunology, Signal Transduction | |
| 5774 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction