missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN13/PTPL1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94207-0.1ml
This item is not returnable.
View return policy
Description
PTPN13/PTPL1 Polyclonal antibody specifically detects PTPN13/PTPL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PTPN13/PTPL1 | |
| Polyclonal | |
| Western Blot 1:2000 - 1:6000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| hPTPE1, PNP1, protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associatedphosphatase), protein-tyrosine phosphatase 1, Fas-associated, PTP1EDKFZp686J1497, PTP-BASEC 3.1.3.48, PTP-E1, PTPL1nonreceptor type 13, tyrosine-protein phosphatase non-receptor type 13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-500 of human PTPN13 (NP_542416.1). QDENYFKDILSDNSGREDSENTFSPYQFKTSGPEKKPIPGIDVLSKKKIWASSMDLLCTADRDFSSGETATYRRCHPEAVTVRTSTTPRKKEARYSDGSIALDIFGPQKMDPIYHTRELPTSSAISSALDRIRERQKKLQVLREAMNVEEPVRRYKTYHGDVFSTSSESPSIISSESDFRQVRRSEASKRFESSSGLPGVDETLSQGQSQRPSRQYETPFEGNLINQEIMLKRQEEELMQLQAKMALRQSR | |
| 0.1 mL | |
| Protein Phosphatase | |
| 5783 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction