missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPMT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00056-100ul
This item is not returnable.
View return policy
Description
PTPMT1 Polyclonal antibody specifically detects PTPMT1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| PTPMT1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| DUSP23, EC 3.1.3.16, EC 3.1.3.48, FLJ46081, MOSPNB4 apoptosis/differentiation related protein, Phosphoinositide lipid phosphatase, PLIPprotein-tyrosine phosphatase mitochondrial 1, protein tyrosine phosphatase, mitochondrial 1, PTEN-like phosphatase | |
| Produced in rabbits immunized with purified, recombinant Human PTPMT1 (Uniprot#: Q8WUK0-1; Lys28-Thr201) | |
| 100 μg | |
| Protein Phosphatase | |
| 114971 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction