missing translation for 'onlineSavingsMsg'
Learn More

PTH Antibody (3H9), DyLight 550 - N-terminal, Novus Biologicals™

Product Code. 18772081 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18772081 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18772081 Supplier Novus Biologicals Supplier No. NBP254320R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTH Monoclonal antibody specifically detects PTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTH
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3H9
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1
Host Species Mouse
Immunogen A synthetic peptide around aa 1-34 of human mature-PTH-polypeptide (exact sequence is proprietary) (Uniprot: P01270)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, BiologicAll speciesy Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5741
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.