missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTER Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77355-0.025mg
This item is not returnable.
View return policy
Description
PTER Polyclonal specifically detects PTER in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PTER | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EC 3.1, hPHRP, Parathion hydrolase-related protein, phosphotriesterase related, phosphotriesterase-related protein, resiniferatoxin-binding, phosphotriesterase-related, RPR-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9317 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| CAH73146 | |
| PTER | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human PTER. The immunogen is located within amino acids 250 - 300 of PTER. Amino Acid Squence: PDIDMPDDNKRIRRVR | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction