missing translation for 'onlineSavingsMsg'
Learn More

PTEN Antibody (CL11722) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30300911 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30300911 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30300911 Supplier Novus Biologicals Supplier No. NBP321200JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTEN Monoclonal antibody specifically detects PTEN in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTEN
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL11722
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias BZS, EC 3.1.3.16,10q23del, EC 3.1.3.48, EC 3.1.3.67, GLM2, MMAC1 phosphatase and tensin homolog deleted on chromosome 10, MMAC1MGC11227, Mutated in multiple advanced cancers 1, phosphatase and tensin homologDEC, phosphatidylinositol-34,5-trisphosphate 3-phosphatase and dual-specificityprotein phosphatase PTEN, PTEN1, TEP1MHAM
Host Species Mouse
Immunogen This antibody was developed using a synthetic peptide derived from P60484, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Biology, Lipid and Metabolism, Metastasis, mTOR Pathway, Phospho Specific, Protein Phosphatase, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5728
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.