missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTCD3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00€ - 470.00€
Specifications
| Antigen | PTCD3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663481
|
Novus Biologicals
NBP2-94697-0.02ml |
0.02 mL |
188.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686180
|
Novus Biologicals
NBP2-94697-0.1ml |
0.1 mL |
470.00€
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PTCD3 Polyclonal antibody specifically detects PTCD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| PTCD3 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 55037 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp666K071, FLJ20758, Pentatricopeptide repeat domain 3, pentatricopeptide repeat-containing protein 3, mitochondrial, Transformation-related gene 15 protein, TRG-15 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-689 of human PTCD3 (NP_060422.4). PDQHRNFYYSKFFDLICLMEQIDVTLKWYEDLIPSAYFPHSQTMIHLLQALDVANRLEVIPKIWKDSKEYGHTFRSDLREEILMLMARDKHPPELQVAFADCAADIKSAYESQPIRQTAQDWPATSLNCIAILFLRAGRTQEAWKMLGLFRKHNKIPRSELLNELMDSAKVSNSPSQAIEVVELASAFSLPICEGLTQRVMSDFAINQEQKEALSNLTALTSDSDTDSSSDSDSDTSEGK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title