missing translation for 'onlineSavingsMsg'
Learn More

PSMG2 Monoclonal Antibody (OTI1E3), TrueMAB™, OriGene

Product Code. 17509083
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17509083 30 μL 30µL
17589073 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17509083 Supplier Origene Technologies Supplier No. TA808142S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PSMG2 Monoclonal antibody specifically detects PSMG2 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen PSMG2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SNRPB2
Gene Accession No. P08579
Gene Alias 1700017I17Rik; AW545363; CD40 ligand-activated specific transcript 3; CLAST3; HCCA3; HDCMC29P; hepatocellular carcinoma susceptibility protein; hepatocellular carcinoma-susceptibility protein 3; HSPC260; HsT1707; likely ortholog of mouse CD40 ligand-activated specific transcript 3 (Clast3); MDS003; PAC2; PAC-2; proteasome (prosome, macropain) assembly chaperone 2; proteasome a; proteasome assembling chaperone 2; proteasome assembly chaperone 2; PSMG2; TNFSF5IP1; Tumor necrosis factor superfamily member 5-induced protein 1; tumor necrosis factor superfamily, member 5-induced protein 1; x 003 protein
Host Species Mouse
Immunogen Full length human recombinant protein of human SNRPB2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6629
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.