missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD5 Mouse anti-Human, Clone: OTI8B1, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811921
This item is not returnable.
View return policy
Description
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator base.Specifications
| PSMD5 | |
| Monoclonal | |
| Unconjugated | |
| PSMD5 | |
| S5B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5711 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI8B1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16401 | |
| PSMD5 | |
| Human recombit protein fragment corresponding to amino acids 2-140 of human PSMD5 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction