missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14899-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to PSMD4.
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.Specifications
| PSMD4 | |
| Polyclonal | |
| Unconjugated | |
| PSMD4 | |
| AF, AF 1, Antisecretory factor 1, ASF, MCB1, PSMD4, pUB R5, Rpn10, S5A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19185, 5710, 83499 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Flow Cytometry | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35226, P55036 | |
| Psmd4 | |
| PSMD4 Fusion Protein Ag6691 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction