Learn More
PSMB8 Rabbit, anti-Human, CoraLite 594, Polyclonal, Proteintech
Product Code. 18076405 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18076405 20 μL 20µL
18008955 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18076405 Supplier Proteintech Supplier No. CL5941485920UL
Rabbit Polyclonal Antibody
The PSMB8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PSMB8. This antibody recognizes human, mouse antigen. The PSMB8 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Antigen PSMB8
Applications Immunofluorescence
Concentration 1000 μg/mL
Conjugate CoraLite 594
Gene Symbols beta5i, D6S216, D6S216E, LMP7, Low molecular mass protein 7, Macropain subunit C13, Proteasome component C13, Proteasome subunit beta 5i, Proteasome subunit beta type 8, PSMB5i, PSMB8, RING10, Y2
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 5696
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG