missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PSMB5 Polyclonal antibody specifically detects PSMB5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | PSMB5 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | LMPXproteasome beta 5 subunit, Macropain epsilon chain, MB1EC 3.4.25.1, Multicatalytic endopeptidase complex epsilon chain, proteasome (prosome, macropain) subunit, beta type, 5, proteasome catalytic subunit 3, Proteasome chain 6, Proteasome epsilon chain, proteasome subunit beta type-5, Proteasome subunit MB1, Proteasome subunit X, proteasome subunit, beta type, 5, PSX large multifunctional protease X, XMGC104214 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: YRDAYSGGAVNLYHVREDGWIRVSSDNVADLHEKYSGST |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?