missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA/FOLH1/NAALADase I Antibody (FOLH1/2354), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08385AF700
This item is not returnable.
View return policy
Description
PSMA/FOLH1/NAALADase I Monoclonal antibody specifically detects PSMA/FOLH1/NAALADase I in Human, Canine, Equine, Feline samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Specifications
| PSMA/FOLH1/NAALADase I | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase | |
| Recombinant human PSMA/FOLH1/NAALADase I protein fragment (around aa 232-433) (exact sequence is proprietary) (Uniprot: Q04609 ) | |
| 100 μg | |
| Cancer, Prostate Cancer | |
| 2346 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
| FOLH1/2354 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Canine, Equine, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction