missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD-95 Antibody (1H4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26619-100ul
This item is not returnable.
View return policy
Description
PSD-95 Monoclonal antibody specifically detects PSD-95 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| PSD-95 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| 1H4 | |
| Western Blot 1:500-1:5000, ELISA | |
| discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15 | |
| A synthesized peptide derived from Human PSD-95 [UniProt P78352] | |
| 100 μL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
| 1742 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction