missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PS 1145 dihydrochloride

Product Code. p-5545979
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18760779

Brand: Tocris Bioscience™ 4569/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective IKK inhibitor; orally active

PS 1145 dihydrochloride is a selective IκB kinase (IKK) inhibitor (IC50 = 100 nM). Has no effect on PKA, PKC and CKII activity. Inhibits IκB-α phosphorylation and NFκB activation in HeLa cells. Inhibits NFκB-mediated proinflammatory gene expression in human airway epithelial cells. Enhances TNF-α-induced apoptosis in a multiple myeloma cell line. Attenuates LPS-induced TNF-α production in vivo. Orally active.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
Quantity 10 mg
Product Type PS 1145 dihydrochloride
Molecular Weight (g/mol) 395.67
Inhibitors PS 1145 dihydrochloride

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.