missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PRRX1 Monoclonal antibody specifically detects PRRX1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PRRX1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | CL14469 |
| Conjugate | Alexa Fluor 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Homeobox protein PHOX1, paired mesoderm homeo box 1, paired mesoderm homeobox 1 isoform pmx-1b, paired mesoderm homeobox protein 1, paired related homeobox 1, Paired-related homeobox protein 1, PHOX1PRX-1, PMX1PRX1 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from P54821, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?