missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRRG2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93368-0.02ml
This item is not returnable.
View return policy
Description
PRRG2 Polyclonal antibody specifically detects PRRG2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PRRG2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| PRGP2proline-rich Gla (G-carboxglutamic acid) polypeptide 2, proline rich Gla (G-carboxyglutamic acid) 2, Proline-rich gamma-carboxyglutamic acid protein 2, proline-rich Gla (G-carboxyglutamic acid) polypeptide 2, Proline-rich Gla protein 2, TMG2, transmembrane gamma-carboxyglutamic acid protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-179 of human PRRG2 (NP_001303264.1). MSLGPEVFLGPPEAQSFLSSHTRIPRANHWDLELLTPGNLERECLEERCSWEEAREYFEDNTLTERFWESYIYNGKGGRGRVDVASLAVGLTGGILLIVLAGLGAFWYLRWRQHRGQQPCPQEAGLISPLSPLNPLGPPTPLPPPPPPPPGLPTYEQALAASGVHDAPPPPYTSLRRPH | |
| 0.02 mL | |
| Cell Biology | |
| 5639 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction