missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRR5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00€
Specifications
| Antigen | PRR5 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Beschreibung
PRR5 Polyclonal antibody specifically detects PRR5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Spezifikation
| PRR5 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_851850 | |
| 55615 | |
| Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human PRR5. The immunogen is located within amino acids 310 - 360 of PRR5. Amino Acid Squence: PSRLYPTTQPPEQGLDPTR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| FLJ20185, FLJ20185k, proline rich 5 (renal), protein observed with Rictor-1 | |
| PRR5 | |
| IgG | |
| Affinity Purified | |
| PRR5 antibody is predicted to not cross-react with other Protor protein family members. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts