missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRR23A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 624.00€
Specifications
| Antigen | PRR23A |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18256441
|
Novus Biologicals
NBP2-59795 |
100 μL |
624.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667258
|
Novus Biologicals
NBP2-59795-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRR23A Polyclonal specifically detects PRR23A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PRR23A | |
| Polyclonal | |
| Purified | |
| RUO | |
| 729627 | |
| This antibody was developed against a recombinant protein corresponding to the amino acid sequence:CASVPEIAAQEEAYEEDADPEFPEL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Unconjugated | |
| Rabbit | |
| Proline Rich 23A | |
| PRR23A | |
| IgG | |
| Protein A purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title