missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35260-100ul
This item is not returnable.
View return policy
Description
PRPS2 Polyclonal antibody specifically detects PRPS2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PRPS2 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, ELISA | |
| EC 2.7.6.1, Phosphoribosyl pyrophosphate synthase II, phosphoribosyl pyrophosphate synthetase 2, PPRibP, PPRibP synthetase, PRSII, PRS-II, ribose-phosphate diphosphokinase 2, ribose-phosphate pyrophosphokinase 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human PRPS2 (NP_002756.1).,, Sequence:, MPNIVLFSGSSHQDLSQRVADRLGLELGKVVTKKFSNQETSVEIGESVRGEDVYIIQSGCGEINDNLMELLIMINACKIASSSRVTAVIPCFPYARQDKKDKSRAPISAKLVANMLSVAGADHIITMDLHASQIQGFFDIPVDNLYAEPAVLQWIRENIAEWKNCIIVSPDAGGAKRVTSIADRLNVEFALIHKERKKANEVDRMVLVGDVKDRVAILVDDMADTCGTICHAADKLLSAG | |
| 100 μL | |
| Lipid and Metabolism | |
| 5634 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction