missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prothrombin Antibody (9A1), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | Prothrombin |
|---|---|
| Clone | 9A1 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043639
|
Novus Biologicals
NBP3-26594-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043472
|
Novus Biologicals
NBP3-26594-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prothrombin Monoclonal antibody specifically detects Prothrombin in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| Prothrombin | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Coagulation factor II, coagulation factor II (thrombin), EC 3.4.21, EC 3.4.21.5, prothrombin, prothrombin B-chain, PT, serine protease | |
| A synthesized peptide derived from Human Prothrombin [UniProt P00734] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 9A1 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2147 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title