missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Z Antibody (2B4), Novus Biologicals™
Mouse Monoclonal Antibody
203.00€
Specifications
| Antigen | Protein Z |
|---|---|
| Clone | 2B4 |
| Dilution | Western Blot 1: 500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Protein Z Monoclonal specifically detects Protein Z in Human samples. It is validated for Western Blot, ELISA.Specifications
| Protein Z | |
| Western Blot 1: 500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| protein Z, vitamin K-dependent plasma glycoprotein, PZ, vitamin K-dependent protein Z | |
| PROZ | |
| IgG1 | |
| Unpurified | |
| 45 kDa |
| 2B4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 8858 | |
| Purified recombinant fragment of Protein Z expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title