missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Protein Phosphatase inhibitor 1 Antibody (SR3103), Novus Biologicals™

Product Code. 30041806 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041806 100 μL 100µL
30041657 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30041806 Supplier Novus Biologicals™ Supplier No. NBP327703100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Protein Phosphatase inhibitor 1 Monoclonal antibody specifically detects Protein Phosphatase inhibitor 1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein Phosphatase inhibitor 1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone SR3103
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:20000, Flow Cytometry 1:20, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Formulation PBS (pH 7.4), 150mM NaCl, 50% glycerol
Gene Alias I1, I-1, IPP-1, IPP1inhibitor-1, protein phosphatase 1 regulatory subunit 1A, protein phosphatase 1, regulatory (inhibitor) subunit 1A, Protein phosphatase inhibitor 1, protein phosphatase inhibitor-1
Host Species Rabbit
Immunogen A synthesized peptide derived from human Protein Phosphatase inhibitor 1 (Uniprot #: Q13522)
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5502
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.