missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Disulfide Isomerase/P4HB Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41319
This item is not returnable.
View return policy
Description
Protein Disulfide Isomerase/P4HB Polyclonal specifically detects Protein Disulfide Isomerase/P4HB in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Protein Disulfide Isomerase/P4HB | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| P4HB | |
| Antibody was raised against a 17 amino acid peptide near the center of human Protein Disulfide Isomerase/P4HB. The immunogen is located within amino acids 190 - 240 of Protein Disulfide Isomerase/P4HB. Amino Acid Squence: FKKFDEGRNNFEGEVTK | |
| Affinity Purified | |
| RUO | |
| Primary | |
| PDIA1 antibody is human, mouse and rat reactive. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml | |
| Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, thyroid hormone-binding protein p55 | |
| Rabbit | |
| 54 kDa | |
| 0.1 mg | |
| Cellular Markers, ER Markers, Fibroblast Cell Markers, Hypoxia | |
| 5034 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction