Learn More
FTM Technologies™ Protection Prefilters
For the filtration of dust and fine particles in the laboratory. FTM Technologies™ Protection Prefilters are for use with fume hoods.
Brand:  FTM Technologies™ PF2
Additional Details : Weight : 0.12000kg
 View more versions of this product
Product Code. 11758024
  • 50.20€ / Pack of 10
Estimated Shipment: 24-05-2024
Description
  • Protects the active charcoal filter against particles
  • Prevents clogging on the filter surface
Specifications
Specifications
HI8F and HI8V Fume Hood Models