Learn More
FTM Technologies™ Protection Prefilters
For the filtration of dust and fine particles in the laboratory. FTM Technologies™ Protection Prefilters are for use with fume hoods.
50.20€ - 54.05€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11728044
|
FTM Technologies™
PFP1 |
Fume Hoods except for HI8F/HI8V Models |
54.05€
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11758024
|
FTM Technologies™
PF2 |
HI8F and HI8V Fume Hood Models |
50.20€
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
- Protects the active charcoal filter against particles
- Prevents clogging on the filter surface