Learn More
FTM Technologies™ Protection Prefilters
For the filtration of dust and fine particles in the laboratory. FTM Technologies™ Protection Prefilters are for use with fume hoods.
Brand: FTM Technologies™ PFP1
Detalles adicionales : Weight : 0.28800kg
Descripción
- Protects the active charcoal filter against particles
- Prevents clogging on the filter surface
Especificaciones
Fume Hoods except for HI8F/HI8V Models |